Savillex PFA Wide-Mouth Sample Bottles
Safely store your samples in chemically inert PFA jars
  • Constructed of durable and chemically inert Perfluoroalkoxy (PFA)
  • Comfortable grip and effortless handling with four ribbed areas
  • Easily view samples with translucent jar
Seamless wide-mouth jars handle severe corrosives, solvents, sensitive reagents, or staining solutions. Withstand temperatures from ‑328°F to 500°F (‑200°C to 260°C). Knurled closures feature an inner secondary seal for permanent storage.
$146.50USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Material Description
Availability Pricing
Savillex Wide-Mouth Sample Jar, PFA; 360 mL EW-06104-15 Mfr # 100-0360-01
COMPARE
360 Perfluoroalkoxy (PFA) Wide-Mouth Sample Bottle, PFA; 360 mL
$146.50USD / Each