SI Analytics BlueLine Electrodes
No more corrosion and interference from Ag/AgCI ions
  • Silamid® reference system eliminates corrosion and interference from silver/silver chloride ions
  • Platinum diaphragm and free flow of electrolyte ensure excellent stability
Laboratory work has never been easier with these new high-quality electrodes from SI Analytics. Innovative design features a sliding gate that allows easy filling of electrolyte solution and seals it from the solution being measured. BlueLine electrodes feature fast adjustment of measured values and high mechanical stability. Operating range is 0 to 14 pH.
Electrode models 55503-00, -30, and -46 offer superior durability with a cone-shaped electrode tip that is rugged and easy to clean. BNC and DIN fixed cable electrodes include a 3-ft (1-m) cable. Order cable 55503-50 with DIN connector or 55503-55 with BNC connector (sold separately) for use with electrodes with removable cable
$187.28 - $472.56USD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
SI Analytics BlueLine 11 pH, Glass pH Electrode with Plug Head EW-55503-00 Mfr # 285129114(BLUE LINE#11PH)
COMPARE
BlueLine 11 pH, Glass pH Electrode with Removeable Cable
$275.35USD / Each
SI Analytics BlueLine 22 pH, Epoxy pH Electrode with Removeable Cable EW-55503-08 Mfr # 285129225(BLUE LINE#22PH)
COMPARE
BlueLine 22 pH, Epoxy pH Electrode with Removeable Cable
$187.28USD / Each
SI Analytics BlueLine 15 pH, Glass pH/ATC Electrode with 3 ft BNC Cable EW-55503-34 Mfr # 285129155(BLUE LINE#15PH)
COMPARE
BlueLine 15 pH, Glass pH/ATC Electrode with 3 ft BNC Cable
$472.56USD / Each
SI Analytics BlueLine 24 pH, Epoxy pH/ATC Electrode with 3 ft DIN Cable EW-55503-38 Mfr # 285129241(BLUE LINE#24PH)
COMPARE
BlueLine 24 pH, Epoxy pH/ATC Electrode with 3 ft DIN Cable
$413.03USD / Each
SI Analytics BlueLine 25 pH, Epxoy pH Electrode with 3 ft BNC Cable EW-55503-40 Mfr # 285129258(BLUE LINE#25PH)
COMPARE
BlueLine 25 pH, Epxoy pH Electrode with 3 ft BNC Cable
$248.05USD / Each
SI Analytics BlueLine 26 pH, Epoxy pH/ATC Electrode with 3 ft BNC Cable EW-55503-44 Mfr # 285129266(BLUE LINE#26PH)
COMPARE
BlueLine 26 pH, Epoxy pH/ATC Electrode with 3 ft BNC Cable
$405.58USD / Each
SI Analytics BlueLine 15 pH, Glass pH Electrode with 3 ft BNC Cable EW-55503-46 Mfr # 285129171(BLUE LINE#17PH)
COMPARE
BlueLine 15 pH, Glass pH Electrode with 3 ft BNC Cable
$296.44USD / Each
Showing 1- 7 of 7