This product is now discontinued, View Related Products

Scienceware Aero Emergency Eyewash Solution

Provides immediate first aid for eye injuries
  • Effectively rinses impurities and irritants from your eyes
  • Neutralizes acids and alkalis that may enter eyes
  • Offers four year shelf life from date of manufacture
  • Meets all FDA requirements including buffered pH solution and preservative as per 21 CFR part 349
The Aero Emergency Eye Wash combines sterile saline with buffered, pH solution to provide fast, safe, and effective first aid for eye injuries. The all-in-one solution quickly neutralizes acid and corrosive chemicals, returning eyes to normal 7.4 pH levels. Ideal for use in laboratory, classroom, work site, and public facilities.

Compliance: ANSI Z358.1-2009
Call For Price

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Scienceware Aero Eyewash; Buffered, pH Neutral .5oz, 5 bottles/pack EW-12345-68 Mfr # 248820001
COMPARE
Eyewash; Buffered, pH Neutral .5oz, 5 bottles/pack
Call For Price
Scienceware Aero Eyewash; Buffered, pH Neutral 5oz, 3 bottles/pack EW-12345-69 Mfr # 248820005
COMPARE
Eyewash; Buffered, pH Neutral 5oz, 3 bottles/pack
Call For Price
Scienceware Aero Eyewash; Buffered, pH Neutral 16oz, 3 bottles/pack EW-12345-70 Mfr # 248820016
COMPARE
Eyewash; Buffered, pH Neutral 16oz, 3 bottles/pack
Call For Price
Scienceware Aero Eyewash; Buffered, pH Neutral 32oz, 3 bottles/pack EW-12345-71 Mfr # 248820032
COMPARE
Eyewash; Buffered, pH Neutral 32oz, 3 bottles/pack
Call For Price
Scienceware Aero Dual Eyewash Station Buffered pH Neutral 16oz EW-12345-72 Mfr # 248825016
COMPARE
Dual Eyewash Station Buffered pH Neutral 16oz
Call For Price
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat