This product is now discontinued, View Related Products

Scienceware Beta Radiation Shields

Nonskid feet prevent slipping on benchtop
Protect yourself from beta radiation and chemical splashes with these 3/8" (9.5 mm) thick acrylic shields. One-piece construction features rounded corners for easy cleanup. Shield has a large work surface and is angled for comfortable viewing.
Call For Price

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Depth (in) Height (in) Width (in)
Availability Pricing
Scienceware F24976-0001 Beta Radiation Shield, 8" x 12" x 14-3/4" EW-86469-15 Mfr # F24976-0001
COMPARE
8 14 3/4 12
Call For Price
Scienceware F24976-0002 Beta Radiation Shield, 12" x 14" x 20-3/4" EW-86469-16 Mfr # F24976-0002
COMPARE
12 20 3/4 14
Call For Price
Scienceware F24976-0003 Beta Radiation Shield, 12-3/8" x 16" x 20-1/2" EW-86469-17 Mfr # F24976-0003
COMPARE
12 3/8 20 1/2 16
Call For Price
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat