Scienceware Earth-Friendly Sampling Sticks, PLA
Lessen environmental impact
  • FDA compliant sticks are safe for food, drug, cosmetic contact
  • Green alternative to petroleum-based sampling tools
These earth-friendly sampling sticks are durably constructed with 100% PLA (polylactic acid) resin. Whether you are mixing samples, handling capsules, scraping containers, or positioning solid samples for examination, these sampling sticks are up to the task.
PLA is a 100% plant-based material which contributes less greenhouse gasses from source to resin than its petroleum-based counterparts. Using annually renewable resources with little environmental impact, these sticks offer a low carbon footprint, are biodegradable, and can be disposed of through industrial composting, incineration, land fill, or recycling. They are comparable in strength and durability to polystyrene, polypropylene, and other plastic resin sampling tools. Nonsterile.
$69.50 - $279.00USD / Pkg of 100
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Material Overall Length (in) Width (in)
Availability Pricing
Scienceware F36931-0000 Earth-Friendly 1001 Stick, 100 pack EW-06263-85 Mfr # F36931-0000
COMPARE
Polylactic Acid (PLA) 3/4
$69.50USD / Pkg of 100
Scienceware F36931-0001 Earth-Friendly 1001 Stick, 500 case EW-06263-86 Mfr # F36931-0001
COMPARE
Polylactic Acid (PLA) 3/4
$279.00USD / Case of 500
Showing 1- 2 of 2
Need Help with this product?
Call Us At1-847-549-7600
Live chat