Scienceware SPINBAR POLYGON W/RING
Center molded pivot ring minimizes the contact area between the bar and vessel; reducing friction and chattering.
$8.10 - $8.80USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Scienceware F371220040 Polygon Spinbar, Magnetic Stir Bar w/Ring, 40 x 8 mm; 1/Pk EW-04775-13 Mfr # F371220040
COMPARE
Polygon Spinbar, Magnetic Stir Bar w/Ring, 40 x 8 mm; 1/Pk
$8.10USD / Each
Scienceware F371220050 Polygon Spinbar, Magnetic Stir Bar w/Ring, 50 x 8 mm; 1/Pk EW-04775-15 Mfr # F371220050
COMPARE
Polygon Spinbar, Magnetic Stir Bar w/Ring, 50 x 8 mm; 1/Pk
$8.80USD / Each