Scientific Industries MultiPlate Genie® High-Speed Vortex Microplate Shakers
Provides uniform mixing in each microplate well
  • Handles up to 4 microplates
  • Compact and rugged design
  • Fully programmable
  • No need for calibration
  • Ideal of use in cold rooms or incubators
  • Fast and easy loading and unloading of plates saves time
The high-speed vortex microplate shaker offers consistent well-to-well mixing in up to 4 microplates, (handles up to 8 microplates with optional accessory tray). Works with PCR plates, deep well plates, 96-well microplates, and 384-well microplates.
The fully programable unit allows you to adjust the time and speed ideal for your microplate sample mixing applications. Easily set the program for timed, continuous run, or interval mixing. For interval mixing, press the pulse feature on the keypad and set the time on and time off. The combined 1 mm orbit and speed setting provides accurate and reliable results every time.
Ideal for many microplate mixing applications such as PCR preparation, digestions, bead mixing, ELISA, EIA, and colorimetric assay.
$995.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Speed (rpm) Power (VAC) Description
Availability Pricing
Scientific Industries SI-4100 MultiPlate Genie® High-Speed Vortex Microplate Shaker; 120V
New Products
EW-51402-31 Mfr # SI-4100
COMPARE
3000 120 MultiPlate Genie® High-Speed Vortex Microplate Shaker; 120V, North America Plug
$995.00USD / Each
Scientific Industries SI-4101 MultiPlate Genie® High-Speed Vortex Microplate Shaker; 230V, No Plug
New Products
EW-51402-33 Mfr # SI-4101
COMPARE
2700 230 MultiPlate Genie® High-Speed Vortex Microplate Shaker; 230V, No Plug
$995.00USD / Each
Scientific Industries SI-4102 MultiPlate Genie® High-Speed Vortex Microplate Shaker; 230V, EU plug
New Products
EW-51402-34 Mfr # SI-4102
COMPARE
2700 230 MultiPlate Genie® High-Speed Vortex Microplate Shaker; 230V, EU plug
$995.00USD / Each
Scientific Industries SI-4103 MultiPlate Genie® High-Speed Vortex Microplate Shaker; 230V, UK plug
New Products
EW-51402-35 Mfr # SI-4103
COMPARE
2700 230 MultiPlate Genie® High-Speed Vortex Microplate Shaker; 230V, UK plug
$995.00USD / Each