This product is now discontinued, View Related Products
Scotsman Cubed Ice Makers
  • Self-contained units eliminate the need to order a separate ice storage bin
  • Compact size allows convenient placement under standard 40''H laboratory benches
  • Slide-away door provides convenient and easy access to ice
Cubed Ice Makers are constructed of durable rust-free rotocast plastic for greater insulation and reduced melting of ice. Ice cube size is approx 7/8'' x 1/2'' x 1''.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Capacity (lbs) Ice/24 hrs (lbs)
Availability Pricing
Scotsman CU1526A Ice-cube maker with air-cooled condenser, 165 Lbs/day production, 115 VAC EW-55100-01 Mfr # CU1526A
COMPARE
86 150
Call For Price
Scotsman CU1526SW-1A Ice-cube maker with water-cooled condenser, 165 Lbs/day production, 115 VAC EW-55100-02 Mfr # CU1526SW-1A
COMPARE
86 150
Call For Price
Scotsman CUO515GA Ice-cube maker with air-cooled condenser, 84 Lbs/day production, 115 VAC EW-55100-05 Mfr # CUO515GA
COMPARE
24 84