Seametrics Electromagnetic Flow Sensors, PVC
Easy installation in difficult applications
  • Use with 1", 1 1/2", and 2" piping
Insertion electromagnetic flowsensors are designed for use with conductive liquids in pipe. They are highly suitable for difficult applications with changing viscosities and pulsating flows such as air-driven diaphragm pumps. With no moving parts, these meters can be used in “dirty” applications where debris would foul a mechanical meter. These flowmeters have no moving parts, are economical, durable, and very easy to install and maintain.
Applications include measurement and control of conductive fluids, measurement for data reporting in industrial processes, control of chemical metering pumps, and fertigation. A choice of materials (stainless steel, brass, and PVC) allows the meter to adapt to a wide range of temperature, pressure, and corrosive environments. These flowmeters h
$903.50 - $1,141.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Pipe Size Description
Availability Pricing
Seametrics Electromagnetic Flow Sensor for 1" Pipe, PVC, 0.2 to 20 ft/sec EW-33126-00 Mfr # EX810-P
COMPARE
1" Electromagnetic Flow Sensor for 1" Pipe, PVC, 0.2 to 20 ft/sec
$903.50USD / Each
Seametrics Electromagnetic Flow Sensor for 1.5" Pipe, PVC, 0.2 to 20 ft/sec EW-33126-02 Mfr # EX810P
COMPARE
1.5" Electromagnetic Flow Sensor for 1.5" Pipe, PVC, 0.2 to 20 ft/sec
$1,141.00USD / Each