Seepex Variable-Speed Progressing Cavity Pump Heads and Pumps
Excellent chemical resistance, pulseless flow, and low shear
  • Gentle low-shear fluid transfer capable of pumping small particles without damaging product
  • Pumps can handle a variety of polymers and chemicals—ideal for pharmaceutical and water treatment applications
  • Sanitary models approved for 3A sanitary applications and can be cleaned in place
A
Pump heads can be mounted on a NEMA 56C frame motor. Pump heads can also be directly connected or belt driven by an AC or DC motor, gasoline engine (min. 2 hp), or air motor.
B
Pumps are close-coupled to an AC motor with an integral VFD controller. The VFD controller features a “hands-off-auto” switch, providing speeds from 30 to 600 rpm.
C
Pumps feature a ½-hp motor with 230/460 VAC input power. This configuration has a mechanical variable-speed drive that adjusts from 0 to 1000 rpm.
Options are broken up into an industrial and a sanitary categories. Other elastomers are avialble, please contact an Applications Specialts for additional information.
$1,411.50 - $7,034.00USD / Each
21 variations of this product are available.
See All Product Options Below
21 PRODUCT OPTIONS
Compareitem specifications
Max Flow Rate (GPH) Motor Type Wetted Materials
Availability Pricing
Seepex Industrial progressing cavity pump with 56C motor mount, 0.34 - 37 GPH EW-76804-03 Mfr # MDP 003-12 56C-bareshaft
COMPARE
37 56C 316 Stainless Steel, Buna-N
$1,411.50USD / Each
Seepex Industrial progressing cavity pump with 56C motor mount, 0.7 - 78 GPH EW-76804-05 Mfr # MDP 006-12 56C-bareshaft
COMPARE
78 56C 316 Stainless Steel, Buna-N
$1,411.50USD / Each
Seepex Industrial progressing cavity pump with 56C motor mount, 1.23 - 156 GPH EW-76804-07 Mfr # MDP 012-12 56C-bareshaft
COMPARE
156 56C 316 Stainless Steel, Buna-N
$1,416.00USD / Each
Seepex Industrial progressing cavity pump with an AC EPT VFD, 1.23 - 78 GPH EW-76804-13 Mfr # MDP 012-12 - AC EPT VFD
COMPARE
78 Variable-frequency drive 316 Stainless Steel, Buna-N
$2,122.50USD / Each
Seepex Industrial progressing cavity pump with 56C motor mount, 0.05 - 7 GPH EW-76804-30 Mfr # MDS 0005-24 56C-BARESHAF
COMPARE
7 56C 316 Stainless Steel, Buna-N
$1,950.00USD / Each
Seepex Industrial progressing cavity pump with 56C motor mount, 0.18 - 17 GPH, 360 psi EW-76804-31 Mfr # MDS 0015-24 56C-BARESHAF
COMPARE
17 56C 316 Stainless Steel, Buna-N
$1,821.00USD / Each
Seepex Industrial progressing cavity pump with 56C motor mount, 0.7 - 78 GPH, 18 psi EW-76804-33 Mfr # MDS 006-12 56C-BARESHAFT
COMPARE
78 56C 316 Stainless Steel, Buna-N
$1,816.00USD / Each
Seepex Industrial progressing cavity pump with 56C motor mount, 2.5 - 451 GPH, 18 psi EW-76804-36 Mfr # MDS 025-12T 56C-BARESHAF
COMPARE
451 56C 316 Stainless Steel, Buna-N
$1,821.00USD / Each
Seepex Industrial progressing cavity pump with 56C motor mount, 3.7 - 831 GPH EW-76804-37 Mfr # MDS 05-6LT 56C-BARESHAFT
COMPARE
831 56C 316 Stainless Steel, Buna-N
$1,821.00USD / Each
Seepex Industrial progressing cavity pump with an AC VFD drive, 0.05 - 3.4 GPH EW-76804-40 Mfr # MD 0005-24 - AC EPT VFD
COMPARE
3.4 Variable-frequency drive 316 Stainless Steel, Buna-N
$2,728.50USD / Each
Seepex Industrial progressing cavity pump with an AC VFD drive, 0.18 - 8.5 GPH EW-76804-41 Mfr # MD 0015-24 - AC EPT VFD
COMPARE
8.5 Variable-frequency drive 316 Stainless Steel, Buna-N
$2,951.50USD / Each
Seepex Industrial progressing cavity pump with an AC VFD drive, 0.34 - 8.5 GPH EW-76804-42 Mfr # MD 003-12 - AC EPT VFD
COMPARE
8.5 Variable-frequency drive 316 Stainless Steel, Buna-N
$2,801.00USD / Each
Seepex Industrial progressing cavity pump with an AC VFD drive, 0.7 - 39 GPH EW-76804-43 Mfr # MD 006-12 - AC EPT VFD
COMPARE
39 Variable-frequency drive 316 Stainless Steel, Buna-N
$2,715.00USD / Each
Seepex Industrial progressing cavity pump with an AC VFD drive, 1.23 - 78 GPH EW-76804-44 Mfr # MD 012-12 - AC EPT VFD
COMPARE
78 Variable-frequency drive 316 Stainless Steel, Buna-N
$2,912.50USD / Each
Seepex Industrial progressing cavity pump with an AC VFD drive, 2.5 - 126 GPH, 90 psi EW-76804-45 Mfr # MD 025-6L - AC EPT VFD
COMPARE
126 Variable-frequency drive 316 Stainless Steel, Buna-N
$2,779.00USD / Each
Seepex Industrial progressing cavity pump with an AC VFD drive, 2.5 - 226 GPH, 18 psi EW-76804-46 Mfr # MD 025-12T - AC EPT VFD
COMPARE
226 Variable-frequency drive 316 Stainless Steel, Buna-N
$2,951.50USD / Each
Seepex Sanitary progressing cavity pump with 56C motor mount, 0.05 - 7 GPH EW-76804-60 Mfr # MDCS 0005-24 56C-BARESHA
COMPARE
7 56C 316 Stainless Stele, Therban®
$5,807.00USD / Each
Seepex Sanitary progressing cavity pump with 56C motor mount, 0.18 - 17 GPH EW-76804-61 Mfr # MDCS 0015-24 56C-BARESHA
COMPARE
17 56C 316 Stainless Stele, Therban®
$5,774.00USD / Each
Seepex Sanitary progressing cavity pump with an AC VFD drive, 0.7 - 39 GPH EW-76804-73 Mfr # MDC 006-12 - AC EPT VFD
COMPARE
39 Variable-frequency drive 316 Stainless Stele, Therban®
$6,765.00USD / Each
Seepex Sanitary progressing cavity pump with an AC VFD drive, 1.23 - 78 GPH EW-76804-74 Mfr # MDC 012-12 - AC EPT VFD
COMPARE
78 Variable-frequency drive 316 Stainless Stele, Therban®
$7,034.00USD / Each
Showing 1 - 20 of 21