Semi-Domed Rugged pH Electrodes
Won’t fail even in tough industrial environments
  • Unique design protects bulb
  • Reduce electrolyte leakage and sample contamination—stand up to viscous samples better than PTFE junctions
  • Extended life in severe applications
  • Graphite-tipped models eliminate electrical noise from your system
Choose from a variety of unique electrode designs for use in tough industrial environments. All models feature BNC pH connectors to match a variety of meters and controllers. Electrodes with temperature compensation feature 100-Ω RTD temperature elements with mini-audio plug connections
These double-junction electrodes have a revolutionary annular ceramic reference junction and a uniform pore size that reduces contamination and clogging.
$263.00 - $283.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Semi-domed pH electrode, PPS housing, 10-ft cable, BNC connector
$263.00USD / Each
COMPARE
Semi-domed pH electrode, PPS housing, 100-Ohm Pt RTD, 10-ft cable, BNC connector
$283.00USD / Each