Seven-Day Timer Pumps
Seven-day timer pumps provide a simple and cost effective metering system for timed applications. Pumps offer eight on/eight off programs per week for maximum adjustability. These pumps feed chemical products on a timed schedule and are commonly used in biocide feed applications. Feed cycle is initiated and controlled by programmable timer.
$616.00 - $632.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (GPH) Max Flow Rate (mL/min) Power (VAC)
Availability Pricing
Pulsafeeder LC13BA-VTC1 Seven-day Timer Pump, 12 GPD, 115 VAC EW-74145-00 Mfr # LC13BA-VTC1
COMPARE
0.51 32 115
$616.00USD / Each
Pulsafeeder LC14BA-VTC1 Seven-day Timer Pump, 22 GPD, 115 VAC EW-74145-10 Mfr # LC14BA-VTC1
COMPARE
0.92 58 115
$616.00USD / Each
Pulsafeeder LC64BA-VTC1 Seven-day Timer Pump, 30 GPD, 115 VAC EW-74145-20 Mfr # LC64BA-VTC1
COMPARE
1.25 79 115
$632.00USD / Each