SGE forte™ Capillary Column for Gas Chromatography, 0.25 mm ID
Offer enhanced endurance without loss of sensitivity
  • SilGuard™ integrated guard column technology provides additional protection by reducing the amount of nonvolatiles entering the analytical column Provides the best possible combination of performance, robustness, reproducibility, low bleed, and inertness Available in a wide variety of phases, IDs, lengths, and film thicknesses
The SGE forte line of GC capillary columns provides the best possible combination of performance, robustness, reproducibility, low bleed, and inertness. It offers a comprehensive range of columns for almost any chromatographic application, including chemical, environmental, food, and fuels.
$329.00 - $1,676.00USD / Each
59 variations of this product are available.
See All Product Options Below
59 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Column Inside Diameter (ID) (mm) Film Thickness (µm) Phase
Availability Pricing
Trajan SGE GC Column, BP1, 100% Dimethyl Polysiloxane, 0.25 mm x 0.1 µm; 15 m length EW-17940-26 Mfr # 054039
COMPARE
0.25 0.1 BP1
$380.00USD / Each
Trajan SGE GC Column, BPX5, 5% Phenyl Polysilphenylene-siloxane, 0.25 mm x 0.25 µm; 15 m length EW-17940-57 Mfr # 054100
COMPARE
0.25 0.25 BPX5
$405.00USD / Each
Trajan SGE GC Column, BPX5, 5% Phenyl Polysilphenylene-siloxane, 0.25 mm x 0.1 µm; 30 m length EW-17940-59 Mfr # 0541011
COMPARE
0.25 0.1 BPX5
$666.00USD / Each
Trajan SGE GC Column, BPX5, 5% Phenyl Polysilphenylene-siloxane, 0.25 mm x 0.5 µm; 30 m length EW-17940-61 Mfr # 0541025
COMPARE
0.25 0.5 BPX5
$666.00USD / Each
Trajan SGE GC Column, BPX5, 5% Phenyl Polysilphenylene-siloxane, 0.25 mm x 1 µm; 15 m length EW-17940-70 Mfr # 054121
COMPARE
0.25 1 BPX5
$421.00USD / Each
Trajan SGE GC Column, BPX5, 5% Phenyl Polysilphenylene-siloxane, 0.25 mm x 1 µm; 60 m length EW-17940-72 Mfr # 054123
COMPARE
0.25 1 BPX5
$1,220.00USD / Each
Trajan SGE GC Column, BP5, 5% Phenyl Methylpolysiloxane, 0.25 mm x 0.25 µm; 60 m length EW-17940-96 Mfr # 054184
COMPARE
0.25 0.25 BP5
$1,028.00USD / Each
Trajan SGE GC Column, BP5, 5% Phenyl Methylpolysiloxane, 0.25 mm x 1 µm; 30 m length EW-17941-03 Mfr # 054203
COMPARE
0.25 1 BP5
$590.00USD / Each
Trajan SGE GC Column, BP20, Polyethylene Glycol, 0.25 mm x 0.5 µm; 30 m length EW-17941-20 Mfr # 054415
COMPARE
0.25 0.5 BP20
$632.00USD / Each
Trajan SGE GC Column, BPX90, 90% Cyanopropyl Polysilphenylene-siloxane, 0.25 mm x 0.25 µm; 30 m length EW-17941-60 Mfr # 054580
COMPARE
0.25 0.25 BPX90
$716.00USD / Each
Trajan SGE GC Column, BPX70, 70% Cyanopropyl Polysilphenylene-siloxane, 0.25 mm x 0.25 µm; 120 m length EW-17941-75 Mfr # 054624
COMPARE
0.25 0.25 BPX70
$1,676.00USD / Each
Trajan SGE GC Column, HT5, 5% Phenyl Polycarborane-siloxane, 0.25 mm x 0.1 µm; 15 m length EW-17941-78 Mfr # 054633
COMPARE
0.25 0.1 HT5
$463.00USD / Each
Trajan SGE GC Column, HT8, 8% Phenyl Polycarborane-siloxane, 0.25 mm x 0.25 µm; 60 m length EW-17941-98 Mfr # 054683
COMPARE
0.25 0.25 HT8
$1,264.00USD / Each
Trajan SGE GC Column, BPX35, 35% Cyanopropyl Polysilphenylene-siloxane, 0.25 mm x 0.25 µm; 30 m length EW-17942-01 Mfr # 054701
COMPARE
0.25 0.25 BPX35
$590.00USD / Each
Trajan SGE GC Column, BPX35, 35% Cyanopropyl Polysilphenylene-siloxane, 0.25 mm x 0.25 µm; 60 m length EW-17942-02 Mfr # 054702
COMPARE
0.25 0.25 BPX35
$1,010.00USD / Each
Trajan SGE GC Column, BPX50, 50% Cyanopropyl Polysilphenylene-siloxane, 0.25 mm x 0.25 µm; 15 m length EW-17942-11 Mfr # 054750
COMPARE
0.25 0.25 BPX50
$463.00USD / Each
Trajan SGE GC Column, BPX50, 50% Cyanopropyl Polysilphenylene-siloxane, 0.25 mm x 0.25 µm; 60 m length EW-17942-13 Mfr # 054752
COMPARE
0.25 0.25 BPX50
$1,180.00USD / Each
Trajan SGE GC Column, SolGel1-MS, 100% Dimethyl Polysiloxane, 0.25 mm x 0.25 µm; 60 m length EW-17942-24 Mfr # 054793
COMPARE
0.25 0.25 SolGel1-MS
$1,054.00USD / Each
Trajan SGE GC Column, SolGel1-MS, 100% Dimethyl Polysiloxane, 0.25 mm x 0.25 µm; 30 m length EW-17942-25 Mfr # 054795
COMPARE
0.25 0.25 SolGel1-MS
$582.00USD / Each
Trajan SGE GC Column, BP624, Cyanopropylphenyl Polysiloxane, 0.25 mm x 1.4 µm; 60 m length EW-17942-51 Mfr # 054842
COMPARE
0.25 1.4 BP624
$1,054.00USD / Each
Showing 1 - 20 of 59