SGE forte™ Capillary Column for Gas Chromatography, 0.53 mm ID
Offer enhanced endurance without loss of sensitivity
  • SilGuard™ integrated guard column technology provides additional protection by reducing the amount of nonvolatiles entering the analytical column Provides the best possible combination of performance, robustness, reproducibility, low bleed, and inertness Available in a wide variety of phases, IDs, lengths, and film thicknesses
The SGE forte line of GC capillary columns provides the best possible combination of performance, robustness, reproducibility, low bleed, and inertness. It offers a comprehensive range of columns for almost any chromatographic application, including chemical, environmental, food, and fuels.
$354.00 - $1,390.00USD / Each
50 variations of this product are available.
See All Product Options Below
50 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Column Inside Diameter (ID) (mm) Film Thickness (µm) Phase
Availability Pricing
Trajan SGE GC Column, BP1, 100% Dimethyl Polysiloxane, 0.53 mm x 1 µm; 30 m length EW-17940-52 Mfr # 054090
COMPARE
0.53 1 BP1
$698.00USD / Each
Trajan SGE GC Column, BP1, 100% Dimethyl Polysiloxane, 0.53 mm x 0.5 µm; 30 m length EW-17940-53 Mfr # 054092
COMPARE
0.53 0.5 BP1
$698.00USD / Each
Trajan SGE GC Column, BPX5, 5% Phenyl Polysilphenylene-siloxane, 0.53 mm x 0.25 µm; 12 m length EW-17940-80 Mfr # 054133
COMPARE
0.53 0.25 BPX5
$421.00USD / Each
Trajan SGE GC Column, BPX5, 5% Phenyl Polysilphenylene-siloxane, 0.53 mm x 0.25 µm; 25 m length EW-17940-81 Mfr # 054134
COMPARE
0.53 0.25 BPX5
$758.00USD / Each
Trajan SGE GC Column, BP5, 5% Phenyl Methylpolysiloxane, 0.53 mm x 1 µm; 30 m length EW-17941-00 Mfr # 054195
COMPARE
0.53 1 BP5
$682.00USD / Each
Trajan SGE GC Column, BP10, 14% Cyanopropylphenyl Polysiloxane, 0.53 mm x 1 µm; 25 m length EW-17941-13 Mfr # 054280
COMPARE
0.53 1 BP10
$674.00USD / Each
Trajan SGE GC Column, BP10, 14% Cyanopropylphenyl Polysiloxane, 0.53 mm x 1 µm; 30 m length EW-17941-14 Mfr # 054283
COMPARE
0.53 1 BP10
$674.00USD / Each
Trajan SGE GC Column, BP20, Polyethylene Glycol, 0.53 mm x 1 µm; 25 m length EW-17941-41 Mfr # 054448
COMPARE
0.53 1 BP20
$758.00USD / Each
Trajan SGE GC Column, BP20, Polyethylene Glycol, 0.53 mm x 2 µm; 25 m length EW-17941-45 Mfr # 054456
COMPARE
0.53 2 BP20
$758.00USD / Each
Trajan SGE GC Column, BP21,Polyethylene Glycol TPA Treated, 0.53 mm x 0.5 µm; 25 m length EW-17941-55 Mfr # 054474
COMPARE
0.53 0.5 BP21
$758.00USD / Each
Trajan SGE GC Column, BP21,Polyethylene Glycol TPA Treated, 0.53 mm x 1 µm; 30 m length EW-17941-57 Mfr # 054478
COMPARE
0.53 1 BP21
$758.00USD / Each
Trajan SGE GC Column, BPX70, 70% Cyanopropyl Polysilphenylene-siloxane, 0.53 mm x 0.5 µm; 30 m length EW-17941-72 Mfr # 054620
COMPARE
0.53 0.5 BPX70
$816.00USD / Each
Trajan SGE GC Column, HT5, 5% Phenyl Polycarborane-siloxane, 0.53 mm x 0.15 µm; 12 m length EW-17941-86 Mfr # 054657
COMPARE
0.53 0.15 HT5
$590.00USD / Each
Trajan SGE GC Column, BPX35, 35% Cyanopropyl Polysilphenylene-siloxane, 0.53 mm x 1 µm; 30 m length EW-17942-07 Mfr # 054737
COMPARE
0.53 1 BPX35
$716.00USD / Each
Trajan SGE GC Column, BPX50, 50% Cyanopropyl Polysilphenylene-siloxane, 0.53 mm x 1 µm; 30 m length EW-17942-15 Mfr # 054772
COMPARE
0.53 1 BPX50
$758.00USD / Each
Trajan SGE GC Column, SolGel Wax, Polyethylene Glycol, 0.53 mm x 0.5 µm; 30 m length EW-17942-18 Mfr # 054786
COMPARE
0.53 0.5 SolGel Wax
$834.00USD / Each
Trajan SGE GC Column, BPX1, 100% Dimethyl Polysiloxane, 0.53 mm x 0.1 µm; 10 m length EW-17942-32 Mfr # 054803
COMPARE
0.53 0.1 BPX1
$379.00USD / Each
Trajan SGE GC Column, BP1, 100% Dimethyl Polysiloxane, 0.53 mm x 5 µm; 60 m length EW-17942-34 Mfr # 054807
COMPARE
0.53 5 BP1
$1,180.00USD / Each
Trajan SGE GC Column, BP1, 100% Dimethyl Polysiloxane, 0.53 mm x 3 µm; 60 m length EW-17942-36 Mfr # 054809
COMPARE
0.53 3 BP1
$1,138.00USD / Each
Trajan SGE GC Column, BP624, Cyanopropylphenyl Polysiloxane, 0.53 mm x 3 µm; 50 m length EW-17942-46 Mfr # 054835
COMPARE
0.53 3 BP624
$1,390.00USD / Each
Showing 1 - 20 of 50