Shake 'n' Bake Shaking Hybridizing Incubators
Adjustable stroke amplitude and speed—vary speeds from 8 to 25 strokes/min
  • Ideal for hybridization of transfer membranes used in Southern and Northern blotting techniques
  • Each removable tray holds one 20 x 20 cm blot
  • PID temperature controller provides rapid heating and temperature stability
Removable shelves accommodate different size containers and heat sealed plastic bags. Maximum shelf load is 2.2 lb (1 kg).
$4,185.00USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC)
Availability Pricing
Boekel Shaking Hybridizing Incubator, 115 VAC, 50/60 Hz, 200 watts EW-39450-10 Mfr # 136400
COMPARE
115
$4,185.00USD / Each