This product is now discontinued, View Related Products
Shimpo Handheld Battery Operated Stroboscope
The TechStrobe is the perfect tool for slow motion or stop action stroboscopic inspection. It is the only palm size unit to include a Phase shift feature. Phase shifting allows the TechStrobe operator to advance or retard the flash timing by degrees of angle, without changing the actual flash rate (FPM) of the TechStrobe.
FEATURES
  • Brightest flash of any strobe in its' class
  • Long run per charge time
  • User adjustable flash rates
  • Buttons for eash x2 and ÷ adjustment
  • Phase shift mode for full inspection of gears and rolls
  • Alphanumeric LCD display with easy user interface
  • One hand operation for right or left handed operators
  • Removable battery with built-in charge electronics
  • External trigger and clock output
  • Includes AC adapter and extra flashtube
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Max Measuring Distance (in) Max RPM Range (rpm) Memory
Availability Pricing
Shimpo BAT-900 Spare Battery Pack for Stroboscope EW-53560-35 Mfr # BAT-900
COMPARE
Call For Price
Shimpo DT-900-PR Techstrobe Professional Kit; 120VAC EW-98807-29 Mfr # DT-900-UP
COMPARE
59000
Call For Price
Shimpo DT-900-UT TechStrobe Traveler Kit; 120VAC EW-98807-30 Mfr # DT-900-UT
COMPARE
59000
Call For Price
Shimpo DT-900-U TechStrobe with Universal AC charger, includes spare flashtube EW-98807-31 Mfr # DT-900-U
COMPARE
59000