Shimpo PT-120 Series Contact Tachometers
Simple-to-use, ergonomic contact tachometers for rotational speed applications
  • Lightweight, ergonomic design minimize user fatigue during extended use
  • Large, backlit LCD for easy viewing in dimly lit areas
  • Automatic recording of minimum, maximum, last value, and up to 96 logged readings
Use these compact digital contact tachometers to measure and analyze rotational speed, distance*, or linear speed. The meter automatically saves maximum, minimum, last recorded values, and up to 96 logged readings. View the measurements on the 5-digit backlit LCD. Select from metric or English unit model.
$95.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max RPM Range (rpm) RPM Accuracy (rpm) Description
Availability Pricing
Shimpo PT-121 Contact Tachometer, rpm, ft/min, feet EW-25303-98 Mfr # PT-121
COMPARE
19.999 ±0.05% + 1 digit Contact Tachometer; rpm, ft/min, feet
$95.00USD / Each
Shimpo PT-122 Contact Tachometer, rpm, m/min, meter EW-25304-93 Mfr # PT-122
COMPARE
19.999 ±0.05% + 1 digit Contact Tachometer; rpm, m/min, meter
$95.00USD / Each