Signet Paddlewheel Flow Sensor
Fluid measurement with paddlewheel sensors
  • Digital (S3L) or 4 to 20mA outputs, or Flow Switch, or Pulse output (multi-function)
  • Allows for up to six sensors to Signet 8900 Controller
  • Low flow capabilities down to 0.1 m/s (0.3 ft/s)
  • Polypropylene or PVDF wetted materials
  • Built-in user interface for on-site configuration
  • Low power and high resolution
$852.00 - $1,150.00USD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Communications Output Pipe Size
Availability Pricing
GF Signet 3-2537-1C-P0 Paddlewheel Flow Sensor, pulse divider/dry contact, PP, 0.5-4" EW-32512-00 Mfr # 3-2537-1C-P0
COMPARE
Pulse divider/dry contact relay ½" to 4" diameter
$852.00USD / Each
GF Signet 3-2537-2C-P0 Paddlewheel Flow Sensor, pulse divider/solid state, PP, 0.5-4" EW-32512-03 Mfr # 3-2537-2C-P0
COMPARE
Pulse divider/solid state relay ½" to 4" diameter
$852.00USD / Each
GF Signet 3-2537-2C-T0 Paddlewheel Flow Sensor, pulse divider/solid state, PVDF, 0.5-4" EW-32512-05 Mfr # 3-2537-2C-T0
COMPARE
Pulse divider/solid state relay ½" to 4" diameter
$1,020.00USD / Each
GF Signet 3-2537-5C-P0 Paddlewheel Flow Sensor, S3L output, PP, 0.5-4" EW-32512-12 Mfr # 3-2537-5C-P0
COMPARE
Digital S3L ½" to 4" diameter
$893.00USD / Each
GF Signet 3-2537-5C-P1 Paddlewheel Flow Sensor, S3L output, PP, 5-8" EW-32512-13 Mfr # 3-2537-5C-P1
COMPARE
Digital S3L 5" to 8" diameter
$936.00USD / Each
GF Signet 3-2537-6C-P0 Paddlewheel Flow Sensor, 4-20 mA output, PP, 0.5-4" EW-32512-15 Mfr # 3-2537-6C-P0
COMPARE
4 to 20 mA ½" to 4" diameter
$963.00USD / Each
GF Signet 3-2537-6C-P1 Paddlewheel Flow Sensor, 4-20 mA output, PP, 5-8" EW-32512-16 Mfr # 3-2537-6C-P1
COMPARE
4 to 20 mA 5" to 8" diameter
$976.00USD / Each
GF Signet 3-2537-6C-T0 Paddlewheel Flow Sensor, 4-20 mA output, PVDF, 0.5-4" EW-32512-17 Mfr # 3-2537-6C-T0
COMPARE
4 to 20 mA ½" to 4" diameter
$1,150.00USD / Each
Showing 1- 8 of 8
Need Help with this product?
Call Us At1-847-549-7600
Live chat