Signet Temperature Sensors
  • RTD sensors are threaded on both ends for submersion in process vessels or in-line applications
  • Use the remote sensor with the field or panel mount transmitter to display temperatures anywhere.
Sensors feature a one-piece body made from PVDF for purity and durability. Available with either a 15-ft or 6'' cable; extend cable up to 200-ft by using two-conductor shielded cable.
$631.50USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Loop Power Supply
Availability Pricing
GF Signet 3-2350-1 Remote Temperature Sensor, 15 ft Cable EW-93310-00 Mfr # 3-2350-1
COMPARE
$631.50USD / Each