This product is now discontinued, View Related Products

Silver Nitrate

Call For Price

12 variations of this product are available.

See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
CAS Number Purity Grade Weight (mL)
Availability Pricing
Ricca Chemical Company 7045-16 Silver Nitrate-Nitric Acid Reagent, for Sulfate Determination (500 mL) EW-86975-58 Mfr # 7045-16
COMPARE
Call For Price
Ricca Chemical Company 6910-32 Silver Nitrate, 0.0192 Normal (0.0192 Molar), 1 mL = 1 mg CN- (1 L) EW-86982-27 Mfr # 6910-32
COMPARE
Call For Price
Ricca Chemical Company 6910-1 Silver Nitrate, 0.0192 Normal (0.0192 Molar), 1 mL = 1 mg CN- (4 L) EW-86987-30 Mfr # 6910-1
COMPARE
Call For Price
Ricca Chemical Company 7045-4 Silver Nitrate-Nitric Acid Reagent, for Sulfate Determination (120 mL) EW-86992-12 Mfr # 7045-4
COMPARE
Call For Price
Fisher Chemical S18125 Silver Nitrate (Crystalline/Certified ACS) (25g) EW-88022-36 Mfr # S18125
COMPARE
7761-88-8
Call For Price
Fisher Chemical S486500 Silver Nitrate (Crystalline/USP) (500g) EW-88022-42 Mfr # S486500
COMPARE
7761-88-8
Call For Price
Fisher Chemical SS68500 Silver Nitrate Solution (1N/Certified) (500mL) EW-88022-44 Mfr # SS68500
COMPARE
7761-88-8
Call For Price
Fisher Chemical SS816500 Silver Nitrate Solution (0.25N) (500mL) EW-88085-53 Mfr # SS816500
COMPARE
7761-88-8
Call For Price
Acros Organics AC197680250 Silver nitrate p.a. (25g) EW-88211-43 Mfr # AC197680250
COMPARE
7761-88-8
Call For Price
Acros Organics AC197681000 Silver nitrate p.a. (100g) EW-88211-44 Mfr # AC197681000
COMPARE
7761-88-8
Call For Price
COMPARE
7761-88-8
Call For Price
COMPARE
7761-88-8
Call For Price
Showing 1- 12 of 12

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat