Silver Sulfate

  • Assay - ≥98.0%
  • Insoluble Matter and AgCl - ≤0.02%
  • Nitrate - ≤0.001%
  • Substances not Precipitated by HCl - ≤0.03%
  • Iron - ≤0.001%
$169.00 - $1,340.00USD / Each

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
CAS Number Weight (g) Description
Availability Pricing
Fisher Chemical S190100 Silver Sulfate (Powder/Certified ACS) (100g) EW-88080-13 Mfr # S190100
COMPARE
10294-26-5 100 Silver Sulfate (Powder/Certified ACS) (100g)
$1,340.00USD / Each
Acros Organics AC209470100 Silver sulfate 99% (10g) EW-88224-10 Mfr # AC209470100
COMPARE
10294-26-5 10 Silver sulfate 99% (10g)
$169.00USD / Each
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat