Simple Wall-Panel Kits
Cost-effective, unassembled kits for your pH metering applications
  • Single- and dual-pump configurations
  • Purchase pump(s) separately or use your existing metering pumps
Any brand of metering pump can be used with these packages—some popular Walchem metering pumps are linked under related products.
Systems include the following components:
Chemical Suction Inlet: 1/2" PVC ball valve
½” PVC fitting
100ML PVC calibration column
1/2" Isolation ball valve
Calibration Inlet: 1/2” PVC ball valve
Pressure gauge with Viton gauge guard
½” NPT PVC coupling
Pipe Supports
1/2" PVC Pipe
Chemical Discharge Outlet: 1/2" PVC Ball Valve
Pump Shelf
$1,990.00 - $2,590.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Differential Pressure (PSI) Max Flow Rate (mL/min) Max Motor RPM (rpm)
Availability Pricing
COMPARE
$1,990.00USD / Each
COMPARE
$2,590.00USD / Each
Showing 1- 2 of 2
Need Help with this product?
Call Us At1-847-549-7600
Live chat