Simpson Analog Safety Volt OHM Meter
  • Certified to EN61010-1 to 600V, Category lll
  • Self contained AC current measurement up to 10 Amps
  • Overload protection available
This new volt-ohm meter is suitable for testing practically all commercial and industrial installations and is rated to Category III-600V applications. Safety has been an important design feature of this meter.
$790.50 - $909.50USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
AC Max (Volts) DC Max (Volts) Description
Availability Pricing
Simpson 260-9S  12397 Analog Safety Volt Ohm Meter with Case EW-20006-04 Mfr # 12397
COMPARE
600 600 Analog Safety Volt Ohm Meter with Case
$790.50USD / Each
Simpson 260-9SP  12398 Analog Multimeter, Safety VOM, Overload Protection, with case EW-20006-05 Mfr # 12398
COMPARE
600 600 Analog Multimeter, Safety VOM, Overload Protection, with case
$909.50USD / Each