Single Head Miniature Diaphragm Pumps
For your laboratory or OEM applications!
These brushless DC motor driven pumps are designed to handle air and gases. The compact design utilizes technologies that allow it to operate harder, hotter, quieter and longer than other existing pump designs. They offer multiple component configurations, allowing them to be used for either vacuum or pressure operation.
$553.00 - $613.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Free-Air Capacity (LPM) Max Vacuum (mm Hg) Description
Availability Pricing
Single Head Miniature Vacuum Pump; 1.5 LPM/12 mmHg-10 psi/12 VDC EW-79600-21 Mfr # L008C-11
COMPARE
1.5 455.7 Single Head Miniature Vacuum Pump; 1.5 LPM/12 mmHg-10 psi/12 VDC
$573.50USD / Each
Parker Single Head Miniature Vacuum Pump; 1.5 LPM/12 mmHg-10 psi/24 VDC EW-79600-22 Mfr # L045B-11
COMPARE
1.5 455.7 Single Head Miniature Vacuum Pump; 1.5 LPM/12 mmHg-10 psi/24 VDC
$613.00USD / Each
Parker Single Head Miniature Vacuum Pump; 2.5 LPM/16 mmHg-20 psi/12 VDC EW-79600-23 Mfr # H004C-11
COMPARE
2.5 354 Single Head Miniature Vacuum Pump; 2.5 LPM/16 mmHg-20 psi/12 VDC
$553.00USD / Each
Parker Single Head Miniature Vacuum Pump; 2.5 LPM/16 mmHg-20 psi/24 VDC EW-79600-24 Mfr # H050D-11
COMPARE
2.5 354 Single Head Miniature Vacuum Pump; 2.5 LPM/16 mmHg-20 psi/24 VDC
$575.00USD / Each