Single Open-End Absolute-Rated Cartridge Systems
A collection to fit a wide range of critical applications
  • Absolute-rated for use in many regulated processes
  • Meets a wide range of industry and government standards
These SOE cartridges have a 222 O-ring configuration to create a tight seal and prevent influent bypass. They are autoclavable to 250°>F (121°>C) and in-line sterilizable to 257°>F (125°>C) for up to 30-minute cycles; tolerant to an accumulated exposure of 10 hours. The filters may also be sanitized with compatible chemical agents. All cartridges are suitable to 180°>F (82°>C) at 10 psid. Use a SOE cartridge-sump adapter kit (sold separately) to secure cartridges within housings.
The cartridges comply with US CFR Title 21 guidelines for repeated food contact, comply with USP Class VI-121°C Plastics guidelines and pass the MEM Elution Cytotoxicity Test. Aqueous extracts contain less than 0.25 EU/mL and typically exhibit low levels of non-volatile residues.
$85.50 - $442.00USD / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Filter Length (in) Filter Media Micron Rating
Availability Pricing
COMPARE
10 Nylon 0.2
$231.00USD / Each
COMPARE
10 Nylon 0.45
$224.00USD / Each
COMPARE
10 Glass fiber 3
$124.00USD / Each
COMPARE
10 Polypropylene 1
$125.00USD / Each
COMPARE
10 Polypropylene 3
$114.00USD / Each
COMPARE
10 Polypropylene 5
$107.00USD / Each
COMPARE
10 Polypropylene 10
$85.50USD / Each
COMPARE
10 PTFE 0.1
$442.00USD / Each
COMPARE
10 PTFE 0.2
$383.00USD / Each
COMPARE
10 PTFE 0.45
$353.00USD / Each
COMPARE
10 Polyethersulfone (PES) 0.2
$239.00USD / Each
Showing 1- 11 of 11