Single-Stage Air Compressors with Storage Tanks

Economical compressors come in a variety of styles for numerous applications
  • Tank styles include: hot-dog, twin-stack, pancake, portable, and vertical
Direct Drive Series features both oilless (07122-00, -05, -10, -15, -40), and oil-lubricated (07122-45) compressors. These lightweight and compact compressors feature die-cast aluminum construction with a cast-iron sleeve and thermal overload protection.
Contractor Series has constant speed operation and a manual reset for overload protection. Splash-lubricated construction with cast-iron sleeve affords a longer compressor life.
Professional Series units are made of 100% cast iron and feature splash-lubricated construction that provides less friction for a longer compressor life. Overload protection is provided through a manual reset.
$236.20 - $693.00USD / Each

6 variations of this product are available.

See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Free Air Capacity (CFM) Power (VAC) Tank Size (Gal)
Availability Pricing
Puma PK2008 Professional Air Compressor, 3.7 cfm, 11 gal Vertical Tank; 115/230V EW-07121-01 Mfr # PK2011VP
COMPARE
3.7 115 8
$395.40USD / Each
Puma PK5020 Professional Air Compressor, 7.1 cfm, 20 gal Tank; 115/230V EW-07121-05 Mfr # PK5020
COMPARE
7.1 115 20
$693.00USD / Each
Puma LA5706 Direct drive air compressor, 3.0 cfm, hotdog tank, 115 VAC EW-07122-00 Mfr # LA5706
COMPARE
3 115 1.5
$236.20USD / Each
Puma PK2008 Direct-Drive Air Compressor, 3.0 cfm, 2 gal Twin-Stack Tank; 115V EW-07122-05 Mfr # LA5721
COMPARE
3 115 2
$245.60USD / Each
Puma LA9021 Direct-Drive Air Compressor, 4.8 cfm, 2 gal Twin-Stack Tank; 115V EW-07122-10 Mfr # LA9021
COMPARE
4.8 115 2
$311.10USD / Each
Puma DP2004V Contractor Air Compressor, 6.0 cfm, 4 gal Twin-Stack Tank; 115V EW-07124-01 Mfr # DP2022S
COMPARE
6 115 4
$382.90USD / Each
Showing 1- 6 of 6

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat