This product is now discontinued, View Related Products

Ricca Chemical Sodium Acetate, 0.200 Molar (M/5)

Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
Call For Price

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
CAS Number Purity Grade Volume (mL)
Availability Pricing
Ricca Chemical Company 7129-16 Sodium Acetate, 0.200 Molar (M/5) (500 mL) EW-86975-65 Mfr # 7129-16
COMPARE
Includes 6131-90-4 and 7732-18-5 Components all ACS 500
Call For Price
Ricca Chemical Company 7129-32 Sodium Acetate, 0.200 Molar (M/5) (32 oz) EW-86982-48 Mfr # 7129-32
COMPARE
Includes 6131-90-4 and 7732-18-5 Components all ACS 1000
Call For Price
Ricca Chemical Company 7129-1 Sodium Acetate, 0.200 Molar (M/5) (4 L) (Liquid) EW-86987-47 Mfr # 7129-1
COMPARE
Includes 6131-90-4 and 7732-18-5 Components all ACS 4000
Call For Price
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat