Sodium Chloride–Tris–EDTA (STE) Buffer, 10X 26256

  • pH of 1X solution at 25°C - 7.9 to 8.1
  • DNase - Not detected
  • RNase - Not detected
  • Protease - Not detected
$84.00USD / Each

1 variations of this product are available.

See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
CAS Number Purity Grade Weight (mL)
Availability Pricing
Fisher BioReagents BP24791 Sodium Chloride-Tris-EDTA (STE) Buffer, 10X (pH 8.0) (1 liter) EW-88059-48 Mfr # BP24791
COMPARE
77861 (Tris Base) 60004 (EDTA) 7647010 (HCl) 7647145 (NaCl)
$84.00USD / Each
Showing 1- 1 of 1

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat