Sodium meta-Silicate Nonahydrate

  • Chloride - ≤0.020%
  • Sulfate - ≤0.010%
  • Iron - ≤0.005%
  • Heavy Metals (as Pb) - ≤0.001%
$283.00USD / Each

1 variations of this product are available.

See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
CAS Number Purity Grade Weight (mL)
Availability Pricing
Fisher Chemical S408500 Sodium meta-Silicate Nonahydrate (Cryst./Certified) (500g) EW-88081-58 Mfr # S408500
COMPARE
13517-24-3 and 6834-92-0 Certified
$283.00USD / Each
Showing 1- 1 of 1

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat