Sodium Nitrite 23478

$46.50 - $459.00USD / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
CAS Number Weight (g) Description
Availability Pricing
Fisher Chemical S347500 Sodium Nitrite (Crystalline/Certified ACS) (500g) EW-88025-02 Mfr # S347500
COMPARE
7632-00-0 500 Sodium Nitrite (Crystalline/Certified ACS) (500g)
$188.00USD / Each
Acros Organics Sodium nitrite, reagent ACS (2kg) CAS 7632-00-0 EW-88362-89 Mfr # AC424350020
COMPARE
7632-00-0 2000 Sodium nitrite, reagent ACS (2kg) CAS 7632-00-0
$459.00USD / Each
COMPARE
7631-99-4 1000 Sodium Nitrite, ACS Grade, 1000 g
$48.50USD / Each
COMPARE
7631-99-4 2500 Sodium Nitrite, ACS Grade, 2.5 K
$68.50USD / Each
COMPARE
7631-99-4 500 Sodium Nitrite, ACS Grade, 500 g
$46.50USD / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat