Sodium Nitrite, 5% (w/w) Aqueous Solution

$27.00 - $44.50USD / Each

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
CAS Number Form Volume (mL)
Availability Pricing
Ricca Chemical Company 7497-16 Sodium Nitrite, 5% (w/w) Aqueous Solution (500 mL) EW-86976-12 Mfr # 7497-16
COMPARE
Includes 7632-00-0 and 7732-18-5 Liquid 500
$27.00USD / Each
Ricca Chemical Company 7497-32 Sodium Nitrite, 5% (w/w) Aqueous Solution (32 oz) EW-86983-15 Mfr # 7497-32
COMPARE
Includes 7632-00-0 and 7732-18-5 Liquid 1000
$44.50USD / Each
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat