This product is now discontinued, View Related Products

Sodium Phosphate Monobasic Dihydrate

  • Assay (anhydrous basis) - ≥99.0%
  • Loss on Drying at 105°C - 21-25%
  • Arsenic - ≤0.5ppm
  • Chloride - ≤5ppm
  • Heavy Metals (as Pb) - ≤0.001%
  • Insoluble Matter, Calcium and NH4OH Precipitate - ≤0.01%
  • Iron - ≤0.001%
  • Nitrogen Compounds (as N) - ≤0.001%
  • pH of a 5% Solution at 25°C - 4.1 to 4.5
  • Sulfate - ≤0.003%
Call For Price

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
CAS Number Purity Grade Weight (mL)
Availability Pricing
Fisher Chemical S3813 Sodium Phosphate Monobasic Dihydrate (Cryst./Certified) (3kg) EW-88081-23 Mfr # S3813
COMPARE
13472-35-0
Call For Price
Fisher Chemical S381500 Sodium Phosphate Monobasic Dihydrate (Cryst./Certified) (500g) EW-88081-24 Mfr # S381500
COMPARE
13472-35-0 Certified
Call For Price
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat