This product is now discontinued, View Related Products

Sodium

  • Chloride - ≤0.002%
  • Nitrogen Compounds (as N) - To pass test (about 0.003%)
  • Phosphate - ≤5ppm
  • Sulfate - To pass test (about 0.002%)
  • Heavy Metals (as Pb) - ≤5ppm
  • Iron - ≤0.001%
Call For Price

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
CAS Number Purity Grade Weight (mL)
Availability Pricing
Fisher Chemical S1351LB Sodium (Certified ACS) (1 lb.) EW-88080-02 Mfr # S1351LB
COMPARE
7440-23-5
Call For Price
Fisher Chemical S2061LB Sodium (1 lb.) 8808017 EW-88080-17 Mfr # S2061LB
COMPARE
7440-23-5
Call For Price
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat