John Guest Acetal Push-To-Connect Soft Tube Supports
No need to buy two fitting halves
  • Meet NSF and FDA requirements for food-grade applications
  • Quick-disconnect convenience for a wide variety of tubing material—including soft metals!
A single fitting offers secure connections and quick disconnection—no need to purchase two fitting halves. Simply push tubing past the O-ring to the internal tube stop—steel teeth on the collet grip tubing for leakproof connections. To disconnect, press collet against fitting body to disengage steel teeth—tubing slides right out.
Fittings work with semirigid plastic (polypropylene, polyethylene, nylon) and soft metal (brass, copper, mild steel) tubing. Use to connect flexible tubing such as Tygon® or silicone.
$3.70 - $7.60USD / Pkg of 10
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 2) (in) Fitting Material Push-to-Connect Tube OD (port 1) (in)
Availability Pricing
John Guest Push-To-Connect Fitting, Soft Tube Support, Acetal, 3/8" OD x 5/16" OD; 10/Pk
Buy More Save More
EW-34007-38 Mfr # TSI312S
COMPARE
5/16 Acetal 3/8
$4.30USD / Pkg of 10
John Guest Push-To-Connect Fitting, Soft Tube Support, Acetal, 1/2" OD x 3/8" OD; 10/Pk
Buy More Save More
EW-34007-39 Mfr # TSI375S
COMPARE
3/8 Acetal 1/2
$7.60USD / Pkg of 10
John Guest Push-To-Connect Fitting, Soft Tube Support, Acetal, 5/16" OD x 1/4" OD; 10/Pk
Buy More Save More
EW-34007-40 Mfr # TSI250S
COMPARE
1/4 Acetal 5/16
$3.70USD / Pkg of 10