SoftCIDE Hand Soap
Use this antibacterial soap to thoroughly clean your hands. The SOFTCIDE soap contains chloroxylenol, which works effectively against gram-positive and gram-negative bacteria, yeast, and fungi. Soap is pH balanced and specially formulated to be nonirritating to your skin—even if you use it more than 40 times a day! Unscented and nonstaining.
$248.00 - $292.00USD / Case of 4
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Format
Availability Pricing
Soft CIDE 21032-06-SGD Hand Soap, 32 oz bottles, 6/cs EW-17791-00 Mfr # 21032-06-001
COMPARE
Pump
$248.00USD / Case of 6
Soft CIDE 21128-04-SGD Hand Soap Refills, Gallon bottles, 4/cs EW-17791-50 Mfr # 21128-04-001
COMPARE
Pump
$292.00USD / Case of 4
Showing 1- 2 of 2
Need Help with this product?
Call Us At1-847-549-7600
Live chat