SoftGUARD Hand Cream

Nongreasy formula absorbs immediately
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.

Relieves irritation, prevent chapping, and protect your hands from frequent washing, solvents, and powdered gloves. Unscented
$94.50 - $132.00USD / Case of 12

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
SoftGUARD 12003-12-SGD Hand Cream, 3 oz Tube; 12 Tubes/Case EW-17790-00 Mfr # 12003-12-001
COMPARE
Hand Cream, 3 oz Tube; 12 Tubes/Case
$94.50USD / Case of 12
SoftGUARD N9340A-ADP Hand Cream, 16 oz Bottle; 6 Bottles/Case EW-17790-10 Mfr # 11016-06-001
COMPARE
Hand Cream, 16 oz Bottle; 6 Bottles/Case
$132.00USD / Case of 6
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat