Masterflex® Solenoid-Operated Pinch Valves for Metric Tubing
Excellent choice for viscous fluids
These solenoid-operated 2- and 3-Way pinch valves are ideal for use with fluids that are difficult for traditional solenoid valves to handle, such as viscous fluids. They are ideal for on/off flow control applications and where frequent changes of wetted parts are necessary. All models are compatible with silicone, C-FLEX®, and PharMed® BPT tubing. The two-way valves are available in both normally closed and normally open configurations. The three-way valves accept two tubes, the bottom tube is normally open while the top is normally closed.
$138.00 - $347.00USD / Each
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
Compareitem specifications
Flow Pattern Pinch Valve Tube Size Power (VDC)
Availability Pricing
COMPARE
Three-Way, One Normally Closed, One Normally Open 2.4 mm OD 12
$153.00USD / Each
COMPARE
Three-Way, One Normally Closed, One Normally Open 2.4 mm OD 24
$152.00USD / Each
COMPARE
Two-Way, Normally Open 10 mm OD 12
$146.00USD / Each
COMPARE
Two-Way, Normally Open 10 mm OD 24
$150.00USD / Each
COMPARE
Two-Way, Normally Open 13 mm OD 12
$297.00USD / Each
COMPARE
Two-Way, Normally Open 13 mm OD 24
$297.00USD / Each
COMPARE
Two-Way, Normally Open 19 mm OD 12
$347.00USD / Each
COMPARE
Two-Way, Normally Open 19 mm OD 24
$344.00USD / Each
COMPARE
Two-Way, Normally Closed 10 mm OD 12
$142.00USD / Each
COMPARE
Two-Way, Normally Closed 10 mm OD 24
$138.00USD / Each
COMPARE
Two-Way, Normally Closed 13 mm OD 12
$288.00USD / Each
COMPARE
Two-Way, Normally Closed 13 mm OD 24
$288.00USD / Each
COMPARE
Two-Way, Normally Closed 19 mm OD 12
$325.00USD / Each
COMPARE
Two-Way, Normally Closed 19 mm OD 24
$323.00USD / Each
Showing 1- 14 of 14