Solvent Recovery Head
Solvent recovery head is used for the condensation and collection of solvents. Use with Kuderna-Danish concentrator 34530-01 or -03. Length: 150 mm.
$690.00USD / Pkg of 1
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Length (cm) Range Resolution
Availability Pricing
Corning 2159-24 Solvent Recovery Head, 1/Pk EW-34511-00 Mfr # 2159-24
COMPARE
$690.00USD / Pkg of 1