Sonicator Microtip Probes
Probes effectively process small samples and attach to the standard 1/2"-dia probe horn. The stepped microtip probe (04711-93) attaches directly to the converter and is ideal for reaching into long, thin tubes.
$427.00 - $471.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Sample Size (mL) Tip Diameter (in)
Availability Pricing
Qsonica 418 Ultrasonic Cell Disruptor Microtip Probes, 1-16" dia EW-04711-89 Mfr # 4417
COMPARE
5 0.063
$471.00USD / Each
Qsonica 419 Ultrasonic Cell Distruptor Microtip Probe 1/8" dia EW-04711-90 Mfr # 4418
COMPARE
15 0.125
$427.00USD / Each
Qsonica 419B Ultrasonic Cell Disruptor Microtip Probes, 1/4" dia EW-04711-92 Mfr # 4420
COMPARE
50 0.25
$427.00USD / Each