This product is now discontinued, View Related Products
SPC Oil Sorbents
  • Soak up spills on land or water!
SPC sorbents are ideal for cleaning up or removing oil from land or water. A single pound of SPC sorbent equals 25 pounds of clay. Each sorbent absorbs up to 25 times its own weight in petroleum products. SPC sorbents are also effective for soaking up petroleum based solvents, paints, vegetable oils, and many other non-water soluble spills. Oil sorbents will not absorb water, will float indefinitely, are wringable, dustless, unaffected by temperature, flame resistant, and impervious to rot and mildew. Booms have a strong mesh netting with metal clips for chaining together and easy retrieval from water. Disposal is easy™each sorbent incinerates to less than 0.2% ash.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Absorption Width (in)
Availability Pricing
SPC100 Heavy weight spill pad EW-86338-00 Mfr # SPC100
COMPARE
36 gal./cs 16
Call For Price
SPC200 Oil Sorbents, Wiper Weight Pad, 16" W x 20" L; 200/cs EW-86338-01 Mfr # SPC200
COMPARE
37 gal./cs 16
Call For Price
SPC810 Boom, 8 inch x 10 foot EW-86338-02 Mfr # SPC810
COMPARE
71 gal./cs 8
Call For Price
SPC510 Boom, 5 inch x 10 foot EW-86338-03 Mfr # SPC510
COMPARE
32 gal./cs 5