Special-Duty Air-Operated Double-Diaphragm Pumps
Unique configurations to meet your demanding needs
  • Santoprene elastomers provide higher abrasion resistance
  • Heavy duty pilot valve simplifies maintenance repairs due to 'dirty' air
  • Stall proof - stops automatically when discharged valve is closed
$1,108.00 - $1,240.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Air Inlet Pressure (PSI) Max Flow Rate (GPM) Max Input VDC (VDC)
Availability Pricing
Warren Rupp G05B1ABTXNSX00 Special-Duty Air-Operated Natural Gas Powered Double-Diaphragm Pump EW-74440-30 Mfr # G05B1ABTXNSX00
COMPARE
100 15
$1,108.00USD / Each
Warren Rupp G1FB1ABTXNSX00 Air-Operated Natural Gas Powered Double-Diaphragm Pump, 1" NPT, Aluminium Body EW-74440-32 Mfr # G1FB1ABTXNSX00
COMPARE
100 45
$1,240.00USD / Each