This product is now discontinued, View Related Products

Specialty Chemicals

Call For Price

50 variations of this product are available.

See All Product Options Below
50 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Fisher Chemical A22325 Periodic Acid Reagent 25g EW-88051-00 Mfr # A22325
COMPARE
Periodic Acid Reagent 25g
Call For Price
Fisher Chemical A289100 Silicotungstic Acid Cert 100g EW-88051-18 Mfr # A289100
COMPARE
Silicotungstic Acid Cert 100g
Call For Price
Fisher Chemical A345500 Aerosol OT Solution, 75%, 500 mL EW-88051-35 Mfr # A345500
COMPARE
Aerosol OT Solution, 75%, 500 mL
Call For Price
Fisher Chemical A349500 Aerosol Ot Solid Anhydrs 500g EW-88051-36 Mfr # A349500
COMPARE
Aerosol Ot Solid Anhydrs 500g
Call For Price
Fisher Chemical A3651 O-phosphoric AC Nf/fcc 1L EW-88051-37 Mfr # A3651
COMPARE
O-phosphoric AC Nf/fcc 1L
Call For Price
Fisher Chemical A657500 Ammonium Carbonate, lumps/NF, 500 g EW-88053-84 Mfr # A657500
COMPARE
Ammonium Carbonate, lumps/NF, 500 g
Call For Price
Fisher Chemical A669C212 Ammon Hydroxide Acs 6x2.5l/cs EW-88053-94 Mfr # A669C212
COMPARE
Ammon Hydroxide Acs 6x2.5l/cs
Call For Price
Fisher Chemical A713500 Amm Meta Vanadate Acs 500gr EW-88054-03 Mfr # A713500
COMPARE
Amm Meta Vanadate Acs 500gr
Call For Price
Fisher BioReagents A867250 Antimony Pot Tartrate Usp 250g EW-88054-27 Mfr # A867250
COMPARE
Antimony Pot Tartrate Usp 250g
Call For Price
Fisher Chemical A998212 Acetonitrile Hplc 2.5L EW-88055-41 Mfr # A998212
COMPARE
Acetonitrile Hplc 2.5L
Call For Price
Fisher BioReagents BP2405SK4 Acetonitrile Hplc Grade EW-88058-72 Mfr # BP2405SK4
COMPARE
Acetonitrile Hplc Grade
Call For Price
Fisher Chemical S47012 Sod Cit Gra Uspfccepbpjp12kg EW-88082-07 Mfr # S47012
COMPARE
Sod Cit Gra Uspfccepbpjp12kg
Call For Price
Fisher Chemical SK154 Kjel-sorb Tm Sol 4liter EW-88084-83 Mfr # SK154
COMPARE
Kjel-sorb Tm Sol 4liter
Call For Price
Acros Organics AC120860050 Humic Acid Sodium Salt Te 5g EW-88118-08 Mfr # AC120860050
COMPARE
Humic Acid Sodium Salt Te 5g
Call For Price
Acros Organics AC121680100 2-hydroxy-5-nitropyridine 10g EW-88119-45 Mfr # AC121680100
COMPARE
2-hydroxy-5-nitropyridine 10g
Call For Price
Acros Organics AC126490250 2-methyl-2-butene 99+% 25ml EW-88126-53 Mfr # AC126490250
COMPARE
2-methyl-2-butene 99+% 25ml
Call For Price
Acros Organics AC129250050 Norcamphor 99% 5g 8813068 EW-88130-68 Mfr # AC129250050
COMPARE
Norcamphor 99% 5g 8813068
Call For Price
Acros Organics AC130180050 Phenethyl Alcohol, 99%, 5 mL EW-88132-21 Mfr # AC130180050
COMPARE
Phenethyl Alcohol, 99%, 5 mL
Call For Price
Acros Organics AC138015000 Tetrachloroethylene, 99%, 500 mL EW-88139-24 Mfr # AC138015000
COMPARE
Tetrachloroethylene, 99%, 500 mL
Call For Price
Acros Organics AC140820020 Vanillin 99% 2g 8814396 EW-88143-96 Mfr # AC140820020
COMPARE
Vanillin 99% 2g 8814396
Call For Price
Showing 1 - 20 of 50

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat