Spectrum Chemical 1,1,2,2-Tetrachloroethane

Quality and delivery you can count on every time!
  • Your source of analytical, research, development, and production-quantity fine chemicals for nearly 40 years
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$84.00 - $1,748.00USD / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Volume (mL) Description
Availability Pricing
Spectrum Chemical 1,1,2,2-Tetrachloroethane; 1 L EW-80216-34 Mfr # T1018-1LTGL
COMPARE
1000 1,1,2,2-Tetrachloroethane; 1 L
$205.00USD / Each
Spectrum Chemical 1,1,2,2-Tetrachloroethane; 20 L EW-80216-35 Mfr # T1018-20LT
COMPARE
20000 1,1,2,2-Tetrachloroethane; 20 L
$1,748.00USD / Each
Spectrum Chemical 1,1,2,2-Tetrachloroethane; 250 mL EW-80216-36 Mfr # T1018-250ML
COMPARE
250 1,1,2,2-Tetrachloroethane; 250 mL
$84.00USD / Each
Spectrum Chemical 1,1,2,2-Tetrachloroethane; 4 L EW-80216-37 Mfr # T1018-4LTGL
COMPARE
4000 1,1,2,2-Tetrachloroethane; 4 L
$688.00USD / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat