Spectrum Chemical 1,10-Phenanthroline Monohydrochloride, Monohydrate

Quality and delivery you can count on every time!
  • Your source of analytical, research, development, and production-quantity fine chemicals for nearly 40 years
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$79.00 - $526.00USD / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Weight (g) Description
Availability Pricing
Spectrum Chemical 1,10-Phenanthroline Monohydrochloride, Monohydrate; 100 g EW-80153-03 Mfr # P1087-100GM
COMPARE
100 1,10-Phenanthroline Monohydrochloride, Monohydrate; 100 g
$526.00USD / Each
Spectrum Chemical 1,10-Phenanthroline Monohydrochloride, Monohydrate; 10 g EW-80153-04 Mfr # P1087-10GM
COMPARE
10 1,10-Phenanthroline Monohydrochloride, Monohydrate; 10 g
$105.00USD / Each
Spectrum Chemical 1,10-Phenanthroline Monohydrochloride, Monohydrate; 25 g EW-80153-05 Mfr # P1087-25GM
COMPARE
25 1,10-Phenanthroline Monohydrochloride, Monohydrate; 25 g
$163.00USD / Each
Spectrum Chemical 1,10-Phenanthroline Monohydrochloride, Monohydrate; 5 g EW-80153-06 Mfr # P1087-5GM
COMPARE
5 1,10-Phenanthroline Monohydrochloride, Monohydrate; 5 g
$79.00USD / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat