Spectrum Chemical 1-Methyl-2-pyrrolidinone

Quality and delivery you can count on every time!
  • Your source of analytical, research, development, and production-quantity fine chemicals for nearly 40 years
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$171.00 - $1,008.00USD / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Purity Grade Volume (mL) Description
Availability Pricing
Spectrum Chemical 1-Methyl-2-pyrrolidinone, BioSolv® Grade; 1 L EW-80040-99 Mfr # BS770-1LT
COMPARE
BioSolv® 1000 1-Methyl-2-pyrrolidinone, BioSolv® Grade; 1 L
$255.25USD / Each
Spectrum Chemical 1-Methyl-2-pyrrolidinone, Reagent ACS Grade; 4 L EW-80128-24 Mfr # M1280-4LTGL
COMPARE
ACS Reagent 4000 1-Methyl-2-pyrrolidinone, Reagent ACS Grade; 4 L
$547.00USD / Each
Spectrum Chemical 1-Methyl-2-pyrrolidinone, Reagent ACS Grade; 500 mL EW-80128-26 Mfr # M1280-500MLGL
COMPARE
ACS Reagent 500 1-Methyl-2-pyrrolidinone, Reagent ACS Grade; 500 mL
$171.00USD / Each
Spectrum Chemical 1-Methyl-2-pyrrolidinone, Electronic/Cleanroom Grade; 1 gal EW-80128-28 Mfr # M1283-1GL
COMPARE
Cleanroom and Electronic 3785 1-Methyl-2-pyrrolidinone, Electronic/Cleanroom Grade; 1 gal
$1,008.00USD / Each
Spectrum Chemical 1-Methyl-2-pyrrolidinone, Anhydrous; 1 L EW-80131-07 Mfr # M1557-1LT
COMPARE
1000 1-Methyl-2-pyrrolidinone, Anhydrous; 1 L
$305.25USD / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat