Spectrum Chemical 2-Mercaptoethanol

Quality and delivery you can count on every time!
  • Your source of analytical, research, development, and production-quantity fine chemicals for nearly 40 years
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$96.50 - $432.00USD / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Volume (mL) Description
Availability Pricing
Spectrum Chemical 2-Mercaptoethanol, Reagent Grade; 100 mL EW-80124-42 Mfr # M1127-100ML
COMPARE
100 2-Mercaptoethanol, Reagent Grade; 100 mL
$96.50USD / Each
Spectrum Chemical 2-Mercaptoethanol, Reagent Grade; 500 mL EW-80124-43 Mfr # M1127-500ML
COMPARE
500 2-Mercaptoethanol, Reagent Grade; 500 mL
$242.00USD / Each
Spectrum Chemical 2-Mercaptoethanol, Electrophoresis Grade; 100 mL EW-80136-65 Mfr # M3030-100ML
COMPARE
100 2-Mercaptoethanol, Electrophoresis Grade; 100 mL
$199.00USD / Each
Spectrum Chemical 2-Mercaptoethanol, Electrophoresis Grade; 25 mL EW-80136-66 Mfr # M3030-25ML
COMPARE
25 2-Mercaptoethanol, Electrophoresis Grade; 25 mL
$163.00USD / Each
Spectrum Chemical 2-Mercaptoethanol, Electrophoresis Grade; 500 mL EW-80136-67 Mfr # M3030-500ML
COMPARE
500 2-Mercaptoethanol, Electrophoresis Grade; 500 mL
$432.00USD / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat