This product is now discontinued, View Related Products

Spectrum Chemical 2-Naphthol

Quality and delivery you can count on every time!
  • Your source of analytical, research, development, and production-quantity fine chemicals for nearly 40 years
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
Call For Price

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Purity Grade Weight (g) Description
Availability Pricing
Spectrum Chemical 2-Naphthol, Reagent Grade; 100 g EW-80141-39 Mfr # N1014-100GM
COMPARE
Reagent 100 2-Naphthol, Reagent Grade; 100 g
Call For Price
Spectrum Chemical 2-Naphthol, Reagent Grade; 500 g EW-80141-40 Mfr # N1014-500GM
COMPARE
Reagent 500 2-Naphthol, Reagent Grade; 500 g
Call For Price
Spectrum Chemical 2-Naphthol, Purified Grade; 12 kg EW-80141-41 Mfr # N1015-12KG
COMPARE
Purified 12000 2-Naphthol, Purified Grade; 12 kg
Call For Price
Spectrum Chemical 2-Naphthol, Purified Grade; 2.5 kg EW-80141-42 Mfr # N1015-2.5KG
COMPARE
Purified 2500 2-Naphthol, Purified Grade; 2.5 kg
Call For Price
Spectrum Chemical 2-Naphthol, Purified Grade; 500 g EW-80141-43 Mfr # N1015-500GM
COMPARE
Purified 500 2-Naphthol, Purified Grade; 500 g
Call For Price
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat