Spectrum Chemical 3-Chloroperoxybenzoic Acid

Quality and delivery you can count on every time!
  • Your source of analytical, research, development, and production-quantity fine chemicals for nearly 40 years
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$92.20 - $5,143.00USD / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Weight (g) Description
Availability Pricing
Spectrum Chemical 3-Chloroperoxybenzoic Acid, Technical Grade; 100 g EW-80045-10 Mfr # C1227-100GM
COMPARE
100 3-Chloroperoxybenzoic Acid, Technical Grade; 100 g
$239.00USD / Each
Spectrum Chemical 3-Chloroperoxybenzoic Acid, Technical Grade; 1 kg EW-80045-11 Mfr # C1227-1KG
COMPARE
1000 3-Chloroperoxybenzoic Acid, Technical Grade; 1 kg
$1,335.50USD / Each
Spectrum Chemical 3-Chloroperoxybenzoic Acid, Technical Grade; 25 g EW-80045-12 Mfr # C1227-25GM
COMPARE
25 3-Chloroperoxybenzoic Acid, Technical Grade; 25 g
$92.20USD / Each
Spectrum Chemical 3-Chloroperoxybenzoic Acid, Technical Grade; 500 g EW-80045-13 Mfr # C1227-500GM
COMPARE
500 3-Chloroperoxybenzoic Acid, Technical Grade; 500 g
$799.25USD / Each
Spectrum Chemical 3-Chloroperoxybenzoic Acid, Technical Grade; 5 kg EW-80045-14 Mfr # C1227-5KG
COMPARE
5000 3-Chloroperoxybenzoic Acid, Technical Grade; 5 kg
$5,143.00USD / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat