This product is now discontinued, View Related Products

Spectrum Chemical 4-Aminobutyric Acid

Quality and delivery you can count on every time!
  • Your source of analytical, research, development, and production-quantity fine chemicals for nearly 40 years
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
Call For Price

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Weight (g) Description
Availability Pricing
Spectrum Chemical 4-Aminobutyric Acid, Reagent Grade; 100 g EW-80022-42 Mfr # AM151-100GM
COMPARE
100 4-Aminobutyric Acid, Reagent Grade; 100 g
Call For Price
Spectrum Chemical 4-Aminobutyric Acid, Reagent Grade; 12 kg EW-80022-43 Mfr # AM151-12KG
COMPARE
12000 4-Aminobutyric Acid, Reagent Grade; 12 kg
Call For Price
Spectrum Chemical 4-Aminobutyric Acid, Reagent Grade; 25 g EW-80022-44 Mfr # AM151-25GM
COMPARE
25 4-Aminobutyric Acid, Reagent Grade; 25 g
Call For Price
Spectrum Chemical 4-Aminobutyric Acid, Reagent Grade; 500 g EW-80022-45 Mfr # AM151-500GM
COMPARE
500 4-Aminobutyric Acid, Reagent Grade; 500 g
Call For Price
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat