Spectrum Chemical Heptane

Quality and delivery you can count on every time!
  • Your source of analytical, research, development, and production-quantity fine chemicals for nearly 40 years
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$124.25 - $1,383.00USD / Each

6 variations of this product are available.

See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Purity Grade Volume (mL) Description
Availability Pricing
Spectrum Chemical Heptane, Reagent Grade; 20 L EW-80098-77 Mfr # H1005-20LTMT
COMPARE
Reagent 20000 Heptane, Reagent Grade; 20 L
$1,383.00USD / Each
Spectrum Chemical Heptane, Reagent Grade; 4 L EW-80098-78 Mfr # H1005-4LTGL
COMPARE
Reagent 4000 Heptane, Reagent Grade; 4 L
$558.00USD / Each
Spectrum Chemical Heptane, Reagent Grade; 500 mL EW-80098-80 Mfr # H1005-500MLGL
COMPARE
Reagent 500 Heptane, Reagent Grade; 500 mL
$189.25USD / Each
Spectrum Chemical Heptane, Practical Grade; 4 L EW-80098-83 Mfr # H1006-4LTGL
COMPARE
Practical 4000 Heptane, Practical Grade; 4 L
$311.00USD / Each
Spectrum Chemical Heptane, Practical Grade; 500 mL EW-80098-84 Mfr # H1006-500MLGL
COMPARE
Practical 500 Heptane, Practical Grade; 500 mL
$124.25USD / Each
Spectrum Chemical Heptane, HPLC Grade; 4 L EW-80108-99 Mfr # HP662-4LT
COMPARE
HPLC 4000 Heptane, HPLC Grade; 4 L
$433.50USD / Each
Showing 1- 6 of 6

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat